Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515283_m1
          See other CIR1 GE Assays ›
          Gene Symbol:
          Cir1
          Gene Name:
          corepressor interacting with RBPJ, 1
          Gene Aliases:
          1700023b02rik, Cir, RGD1309199
          Chromosome Location:
          Chr.3: 59994547 - 60023843 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515283_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cir1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007799.1 NP_001007800.1 8 - 9 592 64
          GenBank mRNA BC085869.1 - 8 - 9 662 7191148 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cir1

          Gene Ontology Categories:

          Function(s)

          transcription, DNA-templated
          mRNA processing
          RNA splicing
          negative regulation of transcription, DNA-templated

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P196856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196856 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P196856 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P196856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846183 - -
          Affymetrix Rat Gene 1.0 ST Array 10796922 - -
          Affymetrix Rat Gene 1.0 ST Array 10846176 - -
          Affymetrix Rat Gene 1.0 ST Array 10846178 - -
          Affymetrix Rat Gene 1.0 ST Array 10846180 - -
          Affymetrix Rat Gene 1.0 ST Array 10846184 - -
          Affymetrix Rat Gene 1.0 ST Array 10846182 - -
          Affymetrix Rat Gene 1.0 ST Array 10846175 - -
          Affymetrix Rat Gene 1.0 ST Array 10846179 - -
          Affymetrix Rat Gene 1.0 ST Array 10846174 - -
          Affymetrix Rat Gene 1.0 ST Array 10846181 - -
          Affymetrix Rat Gene 1.0 ST Array 10846177 - -
          Affymetrix Rat Gene 1.1 ST Array 10846173 - -
          Affymetrix Rat Gene 1.1 ST Array 10796921 - -
          Rat Genome 230 2.0 Array 1374408_PM_at - -
          Rat Genome 230 2.0 Array 1374408_at - -
          Rat Genome 230 2.0 Array 1386229_PM_at - -
          Rat Genome 230 2.0 Array 1386229_at - -
          Affymetrix HT Rat Focus Array 1386229_at - -
          Affymetrix HT Rat Focus Array 1386229_PM_at - -
          Affymetrix HT Rat Focus Array 1374408_at - -
          Affymetrix HT Rat Focus Array 1374408_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300025742.rn.1 TC0300003319.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025741.rn.1 TC0300003319.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025745.rn.1 TC0300003319.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025743.rn.1 TC0300003319.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025744.rn.1 TC0300003319.rn.1 RTA TC
          Rat Expression Set 230 1374408_at - -
          Rat Expression Set 230 1374408_PM_at - -
          Rat Expression Set 230 1386229_PM_at - -
          Rat Expression Set 230 1386229_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846173 - -
          Rat Genome U34 Set AA801048_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21220837 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359605 - -
          RatRef-12 Expression BeadChip ILMN_60755 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline