Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515299_m1
          See other TFPI GE Assays ›
          Gene Symbol:
          Tfpi
          Gene Name:
          tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 71852738 - 71902126 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515299_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfpi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177321.1 NP_001170792.1 5 - 6 513 103
          NM_017200.1 NP_058896.1 4 - 5 441 103
          XM_006234440.2 XP_006234502.1 5 - 6 736 103
          XM_006234441.2 XP_006234503.1 5 - 6 1757 103
          XM_006234444.2 XP_006234506.1 5 - 6 1770 103
          XM_008761980.1 XP_008760202.1 5 - 6 743 103
          XM_008761981.1 XP_008760203.1 5 - 6 507 103
          XM_008761982.1 XP_008760204.1 5 - 6 737 103
          XM_008761983.1 XP_008760205.1 5 - 6 1760 103
          XM_008761984.1 XP_008760206.1 5 - 6 1773 103
          XM_008761985.1 XP_008760207.1 5 - 6 749 103
          XM_008761986.1 XP_008760208.1 5 - 6 743 103
          GenBank mRNA BC087020.1 - 5 - 6 513 7192382 103
          D10926.1 - 4 - 5 441 103
          FQ219876.1 - 2 - 3 147 103
          FQ220339.1 - 2 - 3 141 103
          FQ220655.1 - 2 - 3 238 103
          FQ220711.1 - 2 - 3 238 103
          FQ229364.1 - 2 - 3 170 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003430.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfpi

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119592 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176147 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139731 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011687 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119592 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P726985 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700327072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18598 - -
          Agilent Rat cDNA Microarray (G4105A) 700327072 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846792 - -
          Affymetrix Rat Gene 1.0 ST Array 10846791 - -
          Affymetrix Rat Gene 1.0 ST Array 10846790 - -
          Affymetrix Rat Gene 1.0 ST Array 10846782 - -
          Affymetrix Rat Gene 1.0 ST Array 10846793 - -
          Affymetrix Rat Gene 1.0 ST Array 10846788 - -
          Affymetrix Rat Gene 1.0 ST Array 10846787 - -
          Affymetrix Rat Gene 1.0 ST Array 10846783 - -
          Affymetrix Rat Gene 1.0 ST Array 10846785 - -
          Affymetrix Rat Gene 1.0 ST Array 10846786 - -
          Affymetrix Rat Gene 1.0 ST Array 10846789 - -
          Affymetrix Rat Gene 1.0 ST Array 10846784 - -
          Affymetrix Rat Gene 1.1 ST Array 10846781 - -
          Rat Genome 230 2.0 Array 1368323_PM_at - -
          Rat Genome 230 2.0 Array 1368323_at - -
          Rat Genome 230 2.0 Array 1389780_PM_at - -
          Rat Genome 230 2.0 Array 1389780_at - -
          Affymetrix HT Rat Focus Array 1368323_at - -
          Affymetrix HT Rat Focus Array 1389780_at - -
          Affymetrix HT Rat Focus Array 1368323_PM_at - -
          Affymetrix HT Rat Focus Array 1389780_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300026955.rn.1 TC0300003430.rn.1 RTA TC
          Rat Expression Set 230 1368323_at - -
          Rat Expression Set 230 1389780_PM_at - -
          Rat Expression Set 230 1368323_PM_at - -
          Rat Expression Set 230 1389780_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846781 - -
          Rat Genome U34 Set D10926_s_at - -
          Rat Genome U34 Set rc_AI009783_at - -
          Rat Genome U34 Set rc_AI044095_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21435961 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373066 - -
          RatRef-12 Expression BeadChip ILMN_56355 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline