Search
Search
| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031760.1 | NP_113948.1 | 10 - 11 | 1166 | 71 | |
| GenBank mRNA | AF010597.1 | - | 10 - 11 | 1139 | 71 | |
| U69487.1 | - | 10 - 11 | 1166 | 71 | ||
Set Membership: |
|