Search
Search

Tnfrsf18| Gene Symbol: Tnfrsf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024349.2 | NP_001019520.2 | 1 - 2 | 200 | 75 | |
| XM_008764373.1 | XP_008762595.1 | 1 - 2 | 1455 | 75 | ||
| XM_008764374.1 | XP_008762596.1 | 2 - 3 | 1373 | 75 | ||
| GenBank mRNA | BC088265.1 | - | 1 - 2 | 201 | 7377562 | 75 |
Set Membership: |
|