Search
Search
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239413.2 | XP_006239475.1 | 9 - 10 | 3140 | 89 | |
| XM_006239414.2 | XP_006239476.1 | 9 - 10 | 1638 | 89 | ||
| XM_006239416.1 | XP_006239478.1 | 9 - 10 | 1805 | 89 | ||
| XM_006239417.1 | XP_006239479.1 | 9 - 10 | 1802 | 89 | ||
| XM_006239418.1 | XP_006239480.1 | 8 - 9 | 1737 | 89 | ||
| XM_342975.6 | XP_342976.3 | 9 - 10 | 3143 | 89 | ||
Set Membership: |
|