Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515632_m1
          See other TTLL10 GE Assays ›
          Gene Symbol:
          Ttll10
          Gene Name:
          tubulin tyrosine ligase-like family, member 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 173471020 - 173517785 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttll10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024758.2 NP_001019929.1 6 - 7 620 71
          NM_001170533.1 NP_001164004.1 7 - 8 1042 71
          XM_006239559.2 XP_006239621.1 7 - 8 1202 71
          XM_006239562.2 XP_006239624.1 5 - 6 850 71
          XM_008764356.1 XP_008762578.1 8 - 9 2489 71
          XM_008764357.1 XP_008762579.1 7 - 8 998 71
          GenBank mRNA BC083833.1 - 4 - 5 459 71
          BC097417.1 - 6 - 7 620 7454407 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004224.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ttll10

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyglycylation
          protein-glycine ligase activity
          protein-glycine ligase activity, elongating

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P551001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P551001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P551001 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P551001 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P551001 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882261 - -
          Affymetrix Rat Gene 1.0 ST Array 10882254 - -
          Affymetrix Rat Gene 1.0 ST Array 10882259 - -
          Affymetrix Rat Gene 1.0 ST Array 10882252 - -
          Affymetrix Rat Gene 1.0 ST Array 10882255 - -
          Affymetrix Rat Gene 1.0 ST Array 10882251 - -
          Affymetrix Rat Gene 1.0 ST Array 10882249 - -
          Affymetrix Rat Gene 1.0 ST Array 10882257 - -
          Affymetrix Rat Gene 1.0 ST Array 10882253 - -
          Affymetrix Rat Gene 1.0 ST Array 10882258 - -
          Affymetrix Rat Gene 1.0 ST Array 10882266 - -
          Affymetrix Rat Gene 1.0 ST Array 10882250 - -
          Affymetrix Rat Gene 1.0 ST Array 10882265 - -
          Affymetrix Rat Gene 1.1 ST Array 10882248 - -
          Affymetrix Rat Clariom S Assay PSR0500032272.rn.1 TC0500004224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032271.rn.1 TC0500004224.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032273.rn.1 TC0500004224.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10882248 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21273482 - -
          ABI Rat Genome Survey Microarray 21950758 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359306 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline