Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515736_m1
          See other STK40 GE Assays ›
          Gene Symbol:
          Stk40
          Gene Name:
          serine/threonine kinase 40
          Gene Aliases:
          Lyk4, RGD1307310
          Chromosome Location:
          Chr.5: 144111931 - 144150653 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515736_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stk40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183056.2 NP_898879.2 10 - 11 1253 66
          XM_006238881.2 XP_006238943.1 10 - 11 2963 66
          GenBank mRNA BC100062.1 - 10 - 11 1253 7124923 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stk40

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory system process
          glycogen metabolic process
          protein phosphorylation
          regulation of gene expression
          lung development
          multicellular organism growth
          negative regulation of apoptotic process
          regulation of MAPK cascade
          lung alveolus development
          lung morphogenesis
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1057331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149146 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1057331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1057331 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1057331 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1057331 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P455028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60743 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872011 - -
          Affymetrix Rat Gene 1.0 ST Array 10872004 - -
          Affymetrix Rat Gene 1.0 ST Array 10872013 - -
          Affymetrix Rat Gene 1.0 ST Array 10872012 - -
          Affymetrix Rat Gene 1.0 ST Array 10872006 - -
          Affymetrix Rat Gene 1.0 ST Array 10872007 - -
          Affymetrix Rat Gene 1.0 ST Array 10872010 - -
          Affymetrix Rat Gene 1.0 ST Array 10872005 - -
          Affymetrix Rat Gene 1.0 ST Array 10872008 - -
          Affymetrix Rat Gene 1.0 ST Array 10872009 - -
          Affymetrix Rat Gene 1.0 ST Array 10872014 - -
          Affymetrix Rat Gene 1.1 ST Array 10872003 - -
          Rat Genome 230 2.0 Array 1371943_at - -
          Rat Genome 230 2.0 Array 1371943_PM_at - -
          Affymetrix HT Rat Focus Array 1371943_PM_at - -
          Affymetrix HT Rat Focus Array 1371943_at - -
          Rat Expression Set 230 1371943_at - -
          Rat Expression Set 230 1371943_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872003 - -
          Rat Genome U34 Set rc_AA801260_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366662 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline