Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515792_m1
          See other LUZP1 GE Assays ›
          Gene Symbol:
          Luzp1
          Gene Name:
          leucine zipper protein 1
          Gene Aliases:
          Luzp
          Chromosome Location:
          Chr.5: 154831702 - 154907646 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515792_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Luzp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030830.2 NP_110457.2 4 - 5 3436 66
          XM_006239274.2 XP_006239336.1 3 - 4 3379 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001709.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Luzp1

          Gene Ontology Categories:

          Function(s)

          ventricular septum development
          neural fold bending
          artery development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P372569 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P533778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79241 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P533778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P533778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P372569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P533778 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P533778 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65691 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880690 - -
          Affymetrix Rat Gene 1.0 ST Array 10873017 - -
          Affymetrix Rat Gene 1.0 ST Array 10873014 - -
          Affymetrix Rat Gene 1.0 ST Array 10880688 - -
          Affymetrix Rat Gene 1.0 ST Array 10873016 - -
          Affymetrix Rat Gene 1.0 ST Array 10873013 - -
          Affymetrix Rat Gene 1.0 ST Array 10880687 - -
          Affymetrix Rat Gene 1.0 ST Array 10880689 - -
          Affymetrix Rat Gene 1.0 ST Array 10880686 - -
          Affymetrix Rat Gene 1.1 ST Array 10880685 - -
          Affymetrix Rat Gene 1.1 ST Array 10873012 - -
          Affymetrix Rat Gene 1.1 ST Array 10873015 - -
          Rat Genome 230 2.0 Array 1387003_at - -
          Rat Genome 230 2.0 Array 1387003_PM_at - -
          Affymetrix HT Rat Focus Array 1387003_at - -
          Affymetrix HT Rat Focus Array 1387003_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500012478.rn.1 TC0500001709.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012480.rn.1 TC0500001709.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012479.rn.1 TC0500001709.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012481.rn.1 TC0500001709.rn.1 RTA TC
          Rat Expression Set 230 1387003_PM_at - -
          Rat Expression Set 230 1387003_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873015 - -
          Affymetrix Rat Gene 1.0 ST Array 10873012 - -
          Affymetrix Rat Gene 1.0 ST Array 10880685 - -
          Rat Genome U34 Set rc_AI059074_at - -
          Rat Genome U34 Set rc_AI044249_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21873038 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358314 - -
          RatRef-12 Expression BeadChip ILMN_52416 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline