Search
Search
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145766.2 | NP_665709.2 | 10 - 11 | 1122 | 139 | |
| XM_006239527.2 | XP_006239589.1 | 9 - 10 | 1678 | 139 | ||
| XM_006239528.2 | XP_006239590.1 | 10 - 11 | 1485 | 139 | ||
| XM_006239529.2 | XP_006239591.1 | 10 - 11 | 1406 | 139 | ||
| XM_006239530.1 | XP_006239592.1 | 10 - 11 | 1106 | 139 | ||
| XM_006239531.2 | XP_006239593.1 | 10 - 11 | 1210 | 139 | ||
| XM_006239532.2 | XP_006239594.1 | 5 - 6 | 1554 | 139 | ||
| XM_006239533.1 | XP_006239595.1 | 5 - 6 | 531 | 139 | ||
| GenBank mRNA | DY556124.1 | - | 5 - 6 | 542 | 139 | |
| EX490221.1 | - | 2 - 3 | 212 | 139 | ||
Set Membership: |
|