Search
Search

Pla2g2f| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109587.1 | NP_001103057.1 | 3 - 4 | 429 | 65 | |
| XM_008764289.1 | XP_008762511.1 | 4 - 5 | 854 | 65 | ||
Set Membership: |
|