Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01516203_g1
          See other SH3BGRL3 GE Assays ›
          Gene Symbol:
          Sh3bgrl3
          Gene Name:
          SH3 domain binding glutamate-rich protein like 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 152357270 - 152358643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01516203_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sh3bgrl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106688.1 NP_001100158.1 1 - 2 186 77
          GenBank mRNA BC167002.1 - 1 - 2 111 7443117 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003800.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of actin filament depolymerization
          regulation of actin cytoskeleton organization
          regulation of blood vessel endothelial cell migration
          cell redox homeostasis
          oxidation-reduction process
          electron carrier activity
          protein disulfide oxidoreductase activity
          semaphorin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1000232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000232 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1000232 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000232 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880437 - -
          Affymetrix Rat Gene 1.0 ST Array 10880439 - -
          Affymetrix Rat Gene 1.0 ST Array 10880438 - -
          Affymetrix Rat Gene 1.1 ST Array 10880436 - -
          Rat Genome 230 2.0 Array 1371487_at - -
          Rat Genome 230 2.0 Array 1371487_PM_at - -
          Affymetrix HT Rat Focus Array 1371487_at - -
          Affymetrix HT Rat Focus Array 1371487_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028478.rn.1 TC0500003800.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028476.rn.1 TC0500003800.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028479.rn.1 TC0500003800.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028477.rn.1 TC0500003800.rn.1 RTA TC
          Rat Expression Set 230 1371487_PM_at - -
          Rat Expression Set 230 1371487_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880436 - -
          Rat Genome U34 Set rc_AI172086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21360258 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0