Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01516663_g1
          See other ZBTB17 GE Assays ›
          Gene Symbol:
          Zbtb17
          Gene Name:
          zinc finger and BTB domain containing 17
          Gene Aliases:
          Zfp100
          Chromosome Location:
          Chr.5: 159993799 - 160014899 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01516663_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012105.1 NP_001012105.1 15 - 16 2378 66
          XM_006239303.1 XP_006239365.1 15 - 16 2352 66
          XM_008764253.1 XP_008762475.1 14 - 15 2263 66
          GenBank mRNA BC083577.1 - 15 - 16 2378 7113925 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001822.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb17

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          gastrulation with mouth forming second
          ectoderm development
          positive regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P462010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44180 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462010 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462010 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P462010 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P462010 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80095 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26508 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873751 - -
          Affymetrix Rat Gene 1.0 ST Array 10873744 - -
          Affymetrix Rat Gene 1.0 ST Array 10873737 - -
          Affymetrix Rat Gene 1.0 ST Array 10873752 - -
          Affymetrix Rat Gene 1.0 ST Array 10873746 - -
          Affymetrix Rat Gene 1.0 ST Array 10873739 - -
          Affymetrix Rat Gene 1.0 ST Array 10873741 - -
          Affymetrix Rat Gene 1.0 ST Array 10873743 - -
          Affymetrix Rat Gene 1.0 ST Array 10873745 - -
          Affymetrix Rat Gene 1.0 ST Array 10873740 - -
          Affymetrix Rat Gene 1.0 ST Array 10873747 - -
          Affymetrix Rat Gene 1.0 ST Array 10873753 - -
          Affymetrix Rat Gene 1.0 ST Array 10873748 - -
          Affymetrix Rat Gene 1.0 ST Array 10873738 - -
          Affymetrix Rat Gene 1.0 ST Array 10873750 - -
          Affymetrix Rat Gene 1.0 ST Array 10873742 - -
          Affymetrix Rat Gene 1.0 ST Array 10873749 - -
          Affymetrix Rat Gene 1.1 ST Array 10873736 - -
          Rat Genome 230 2.0 Array 1372880_PM_at - -
          Rat Genome 230 2.0 Array 1372880_at - -
          Affymetrix HT Rat Focus Array 1372880_at - -
          Affymetrix HT Rat Focus Array 1372880_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500013800.rn.1 TC0500001822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013797.rn.1 TC0500001822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013798.rn.1 TC0500001822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013801.rn.1 TC0500001822.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013799.rn.1 TC0500001822.rn.1 RTA TC
          Rat Expression Set 230 1372880_at - -
          Rat Expression Set 230 1372880_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873736 - -
          Rat Genome U34 Set rc_AI104153_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21466530 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376446 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376029 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline