Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01516709_m1
          See other PIK3CD GE Assays ›
          Gene Symbol:
          Pik3cd
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 166602053 - 166628028 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01516709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3cd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108978.1 NP_001102448.1 2 - 3 298 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004085.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          cell surface receptor signaling pathway
          phosphatidylinositol-3-phosphate biosynthetic process
          B cell activation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P956502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72067 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P956502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P956502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P956502 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213363 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881778 - -
          Affymetrix Rat Gene 1.0 ST Array 10881788 - -
          Affymetrix Rat Gene 1.0 ST Array 10881783 - -
          Affymetrix Rat Gene 1.0 ST Array 10881799 - -
          Affymetrix Rat Gene 1.0 ST Array 10881792 - -
          Affymetrix Rat Gene 1.0 ST Array 10881794 - -
          Affymetrix Rat Gene 1.0 ST Array 10881793 - -
          Affymetrix Rat Gene 1.0 ST Array 10881797 - -
          Affymetrix Rat Gene 1.0 ST Array 10881784 - -
          Affymetrix Rat Gene 1.0 ST Array 10881782 - -
          Affymetrix Rat Gene 1.0 ST Array 10881798 - -
          Affymetrix Rat Gene 1.0 ST Array 10881787 - -
          Affymetrix Rat Gene 1.0 ST Array 10881781 - -
          Affymetrix Rat Gene 1.0 ST Array 10881780 - -
          Affymetrix Rat Gene 1.0 ST Array 10881800 - -
          Affymetrix Rat Gene 1.0 ST Array 10881796 - -
          Affymetrix Rat Gene 1.0 ST Array 10881795 - -
          Affymetrix Rat Gene 1.0 ST Array 10881790 - -
          Affymetrix Rat Gene 1.0 ST Array 10881785 - -
          Affymetrix Rat Gene 1.0 ST Array 10881779 - -
          Affymetrix Rat Gene 1.0 ST Array 10881786 - -
          Affymetrix Rat Gene 1.1 ST Array 10881777 - -
          Rat Genome 230 2.0 Array 1393755_PM_at - -
          Rat Genome 230 2.0 Array 1393755_at - -
          Rat Genome 230 2.0 Array 1373528_at - -
          Rat Genome 230 2.0 Array 1373528_PM_at - -
          Affymetrix HT Rat Focus Array 1393755_at - -
          Affymetrix HT Rat Focus Array 1373528_at - -
          Affymetrix HT Rat Focus Array 1393755_PM_at - -
          Affymetrix HT Rat Focus Array 1373528_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500031242.rn.1 TC0500004085.rn.1 RTA TC
          Rat Expression Set 230 1373528_at - -
          Rat Expression Set 230 1373528_PM_at - -
          Rat Expression Set 230 1393755_PM_at - -
          Rat Expression Set 230 1393755_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881777 - -
          Rat Genome U34 Set rc_AA893007_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20860437 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline