Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01517100_g1
          See other SNW1 GE Assays ›
          Gene Symbol:
          Snw1
          Gene Name:
          SNW domain containing 1
          Gene Aliases:
          RGD1561926
          Chromosome Location:
          Chr.6: 111483698 - 111507782 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01517100_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snw1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109279.1 NP_001102749.1 9 - 10 950 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mRNA splicing, via spliceosome
          biological_process
          positive regulation of transforming growth factor beta receptor signaling pathway
          skeletal muscle cell differentiation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation by host of viral transcription
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of mRNA splicing, via spliceosome
          retinoic acid receptor signaling pathway
          regulation of retinoic acid receptor signaling pathway
          positive regulation of neurogenesis
          positive regulation of histone H3-K4 methylation
          regulation of vitamin D receptor signaling pathway
          positive regulation of vitamin D receptor signaling pathway
          cellular response to retinoic acid
          transcription coactivator activity
          transcription corepressor activity
          Notch binding
          protein binding
          nuclear hormone receptor binding
          vitamin D receptor binding
          retinoic acid receptor binding
          poly(A) RNA binding
          SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P204078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891380 - -
          Affymetrix Rat Gene 1.0 ST Array 10891374 - -
          Affymetrix Rat Gene 1.0 ST Array 10891379 - -
          Affymetrix Rat Gene 1.0 ST Array 10891377 - -
          Affymetrix Rat Gene 1.0 ST Array 10891378 - -
          Affymetrix Rat Gene 1.0 ST Array 10891383 - -
          Affymetrix Rat Gene 1.0 ST Array 10891375 - -
          Affymetrix Rat Gene 1.0 ST Array 10891381 - -
          Affymetrix Rat Gene 1.0 ST Array 10891376 - -
          Affymetrix Rat Gene 1.0 ST Array 10891382 - -
          Affymetrix Rat Gene 1.1 ST Array 10891373 - -
          Affymetrix Rat Clariom S Assay PSR0600020726.rn.1 TC0600002917.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10891373 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22188736 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline