Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01517119_m1
          See other SERPINA6 GE Assays ›
          Gene Symbol:
          Serpina6
          Gene Name:
          serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6
          Gene Aliases:
          Cbg
          Chromosome Location:
          Chr.6: 127523948 - 127534178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01517119_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009663.1 NP_001009663.1 4 - 5 1077 79
          GenBank mRNA BC088300.1 - 4 - 5 1077 7323342 79
          FQ209415.1 - 4 - 5 1122 79
          FQ211289.1 - 4 - 5 1092 79
          FQ218294.1 - 4 - 5 1100 79
          FQ218398.1 - 4 - 5 1087 79
          FQ219289.1 - 4 - 5 1152 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003071.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina6

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          glucocorticoid metabolic process
          negative regulation of endopeptidase activity
          molecular_function
          serine-type endopeptidase inhibitor activity
          steroid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P121347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959864 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121347 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121347 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P121347 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121347 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891923 - -
          Affymetrix Rat Gene 1.0 ST Array 10891927 - -
          Affymetrix Rat Gene 1.0 ST Array 10891924 - -
          Affymetrix Rat Gene 1.0 ST Array 10891926 - -
          Affymetrix Rat Gene 1.0 ST Array 10891925 - -
          Affymetrix Rat Gene 1.1 ST Array 10891922 - -
          Rat Genome 230 2.0 Array 1373686_at - -
          Rat Genome 230 2.0 Array 1373686_PM_at - -
          Affymetrix HT Rat Focus Array 1373686_at - -
          Affymetrix HT Rat Focus Array 1373686_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600021862.rn.1 TC0600003071.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600021864.rn.1 TC0600003071.rn.1 RTA TC
          Rat Expression Set 230 1373686_at - -
          Rat Expression Set 230 1373686_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891922 - -
          Rat Genome U34 Set rc_AA893495_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21943534 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359537 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040005 - -
          RatRef-12 Expression BeadChip ILMN_51775 - -
          RatRef-12 Expression BeadChip ILMN_161864 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-snt5b:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline