Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01517232_m1
          See other ZC3H14 GE Assays ›
          Gene Symbol:
          Zc3h14
          Gene Name:
          zinc finger CCCH type containing 14
          Gene Aliases:
          Npuk68
          Chromosome Location:
          Chr.6: 122729837 - 122767462 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01517232_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zc3h14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033951.1 NP_001029123.1 15 - 16 2304 71
          NM_138920.1 NP_620275.1 13 - 14 1911 71
          XM_008764738.1 XP_008762960.1 15 - 16 2338 71
          XM_008764739.1 XP_008762961.1 13 - 14 1945 71
          XM_008764740.1 XP_008762962.1 5 - 6 1666 71
          GenBank mRNA AB032932.1 - 13 - 14 1911 71
          AB097075.1 - 15 - 16 2304 71
          AB097076.1 - 13 - 14 1911 71
          AB097077.1 - 15 - 16 2304 71
          BC087712.1 - 13 - 14 1917 7317608 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001323.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zc3h14

          Gene Ontology Categories:

          Process(es)

          RNA binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P487081 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487081 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487081 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P487081 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501268 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8514 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886319 - -
          Affymetrix Rat Gene 1.0 ST Array 10886306 - -
          Affymetrix Rat Gene 1.0 ST Array 10886310 - -
          Affymetrix Rat Gene 1.0 ST Array 10886317 - -
          Affymetrix Rat Gene 1.0 ST Array 10886302 - -
          Affymetrix Rat Gene 1.0 ST Array 10886318 - -
          Affymetrix Rat Gene 1.0 ST Array 10886307 - -
          Affymetrix Rat Gene 1.0 ST Array 10886299 - -
          Affymetrix Rat Gene 1.0 ST Array 10886316 - -
          Affymetrix Rat Gene 1.0 ST Array 10886300 - -
          Affymetrix Rat Gene 1.0 ST Array 10886309 - -
          Affymetrix Rat Gene 1.0 ST Array 10886321 - -
          Affymetrix Rat Gene 1.0 ST Array 10886313 - -
          Affymetrix Rat Gene 1.0 ST Array 10886304 - -
          Affymetrix Rat Gene 1.0 ST Array 10886314 - -
          Affymetrix Rat Gene 1.0 ST Array 10886320 - -
          Affymetrix Rat Gene 1.0 ST Array 10886303 - -
          Affymetrix Rat Gene 1.0 ST Array 10886301 - -
          Affymetrix Rat Gene 1.0 ST Array 10886315 - -
          Affymetrix Rat Gene 1.0 ST Array 10886308 - -
          Affymetrix Rat Gene 1.1 ST Array 10886298 - -
          Rat Genome 230 2.0 Array 1387921_PM_at - -
          Rat Genome 230 2.0 Array 1387921_at - -
          Rat Genome 230 2.0 Array 1385514_PM_at - -
          Rat Genome 230 2.0 Array 1390297_at - -
          Rat Genome 230 2.0 Array 1385514_at - -
          Rat Genome 230 2.0 Array 1390297_PM_at - -
          Affymetrix HT Rat Focus Array 1390297_PM_at - -
          Affymetrix HT Rat Focus Array 1385514_PM_at - -
          Affymetrix HT Rat Focus Array 1390297_at - -
          Affymetrix HT Rat Focus Array 1387921_PM_at - -
          Affymetrix HT Rat Focus Array 1385514_at - -
          Affymetrix HT Rat Focus Array 1387921_at - -
          Affymetrix Rat Clariom S Assay PSR0600009162.rn.1 TC0600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009164.rn.1 TC0600001323.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009161.rn.1 TC0600001323.rn.1 RTA TC
          Rat Expression Set 230 1390297_PM_at - -
          Rat Expression Set 230 1387921_at - -
          Rat Expression Set 230 1390297_at - -
          Rat Expression Set 230 1387921_PM_at - -
          Rat Expression Set 230 1385514_at - -
          Rat Expression Set 230 1385514_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886298 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21485301 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365742 - -
          RatRef-12 Expression BeadChip ILMN_69787 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline