Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01517603_m1
          See other TC2N GE Assays ›
          Gene Symbol:
          Tc2n
          Gene Name:
          tandem C2 domains, nuclear
          Gene Aliases:
          Mtac2d1, Tac2-N
          Chromosome Location:
          Chr.6: 125437767 - 125601408 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01517603_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tc2n
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025152.1 NP_001020323.1 13 - 14 1647 85
          XM_008764831.1 XP_008763053.1 12 - 13 3310 85
          XM_008764832.1 XP_008763054.1 13 - 14 2959 85
          XM_008764833.1 XP_008763055.1 13 - 14 3206 85
          XM_008764834.1 XP_008763056.1 12 - 13 2180 85
          XM_008764835.1 XP_008763057.1 13 - 14 2211 85
          XM_008764836.1 XP_008763058.1 13 - 14 1829 85
          XM_008764837.1 XP_008763059.1 13 - 14 2174 85
          XM_008764838.1 XP_008763060.1 12 - 13 2143 85
          XM_008764839.1 XP_008763061.1 12 - 13 1709 85
          XM_008764840.1 XP_008763062.1 11 - 12 1576 85
          GenBank mRNA BC088432.1 - 13 - 14 1647 7124658 85
          FQ211170.1 - 13 - 14 1765 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003037.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tc2n

          Gene Ontology Categories:

          Function(s)

          regulation of gene expression
          regulation of blood coagulation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P749222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77133 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P749222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P749222 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P749222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P760599 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P590999 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53472 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39168 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53361 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891775 - -
          Affymetrix Rat Gene 1.0 ST Array 10891767 - -
          Affymetrix Rat Gene 1.0 ST Array 10891772 - -
          Affymetrix Rat Gene 1.0 ST Array 10891771 - -
          Affymetrix Rat Gene 1.0 ST Array 10891768 - -
          Affymetrix Rat Gene 1.0 ST Array 10891770 - -
          Affymetrix Rat Gene 1.0 ST Array 10891776 - -
          Affymetrix Rat Gene 1.0 ST Array 10891778 - -
          Affymetrix Rat Gene 1.0 ST Array 10891773 - -
          Affymetrix Rat Gene 1.0 ST Array 10891766 - -
          Affymetrix Rat Gene 1.0 ST Array 10891779 - -
          Affymetrix Rat Gene 1.0 ST Array 10891769 - -
          Affymetrix Rat Gene 1.0 ST Array 10891777 - -
          Affymetrix Rat Gene 1.1 ST Array 10891765 - -
          Rat Genome 230 2.0 Array 1383606_PM_at - -
          Rat Genome 230 2.0 Array 1383606_at - -
          Affymetrix HT Rat Focus Array 1383606_at - -
          Affymetrix HT Rat Focus Array 1383606_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600021497.rn.1 TC0600003037.rn.1 RTA TC
          Rat Expression Set 230 1383606_PM_at - -
          Rat Expression Set 230 1383606_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891765 - -
          Rat Genome U34 Set rc_AI639417_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21268276 - -
          ABI Rat Genome Survey Microarray 21564167 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374410 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline