Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01517876_g1
          See other IFI27 GE Assays ›
          Gene Symbol:
          Ifi27
          Gene Name:
          interferon, alpha-inducible protein 27
          Gene Aliases:
          IRG1, Ifi27l, Ifi27l1, isg12(a)
          Chromosome Location:
          Chr.6: 127327959 - 127334430 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01517876_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203410.1 NP_981955.1 5 - 6 517 62
          GenBank mRNA BC060548.1 - 5 - 6 517 6919535 62
          FQ219860.1 - 5 - 6 575 62
          FQ220098.1 - 5 - 6 586 62
          FQ226050.1 - 4 - 5 636 62
          FQ226998.1 - 5 - 6 550 62
          FQ230691.1 - 4 - 5 537 62
          FQ233730.1 - 5 - 6 551 62
          FQ233981.1 - 5 - 6 586 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001393.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi27

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to estradiol
          response to cytokine
          regulation of protein export from nucleus
          maternal process involved in female pregnancy
          apoptotic signaling pathway
          RNA polymerase II activating transcription factor binding
          molecular_function
          lamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80552 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148433 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148433 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886579 - -
          Affymetrix Rat Gene 1.0 ST Array 10886574 - -
          Affymetrix Rat Gene 1.0 ST Array 10886578 - -
          Affymetrix Rat Gene 1.0 ST Array 10886576 - -
          Affymetrix Rat Gene 1.0 ST Array 10886575 - -
          Affymetrix Rat Gene 1.0 ST Array 10886580 - -
          Affymetrix Rat Gene 1.0 ST Array 10886581 - -
          Affymetrix Rat Gene 1.1 ST Array 10886573 - -
          Rat Genome 230 2.0 Array 1387770_PM_at - -
          Rat Genome 230 2.0 Array 1387770_at - -
          Affymetrix HT Rat Focus Array 1387770_PM_at - -
          Affymetrix HT Rat Focus Array 1387770_at - -
          Affymetrix Rat Clariom S Assay PSR0600009707.rn.1 TC0600001393.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009709.rn.1 TC0600001393.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009706.rn.1 TC0600001393.rn.1 RTA TC
          Rat Expression Set 230 1387770_PM_at - -
          Rat Expression Set 230 1387770_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886573 - -
          Rat Genome U34 Set rc_AA945204_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21227858 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374967 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650955 - -
          RatRef-12 Expression BeadChip ILMN_48424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline