Search
Search
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240365.2 | XP_006240427.1 | 2 - 3 | 278 | 89 | |
| XM_008764884.1 | XP_008763106.1 | 2 - 3 | 278 | 89 | ||
| XM_008764885.1 | XP_008763107.1 | 2 - 3 | 197 | 89 | ||
| XM_008764886.1 | XP_008763108.1 | 3 - 4 | 433 | 89 | ||
| XM_008764887.1 | XP_008763109.1 | 3 - 4 | 293 | 89 | ||
| XM_008764888.1 | XP_008763110.1 | 2 - 3 | 280 | 89 | ||
Set Membership: |
|