Search
Search
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108053.1 | NP_001101523.1 | 3 - 4 | 410 | 71 | |
| XM_006240477.2 | XP_006240539.1 | 4 - 5 | 750 | 71 | ||
| GenBank mRNA | BC168869.1 | - | 3 - 4 | 327 | 7104604 | 71 |
| FQ211938.1 | - | 3 - 4 | 396 | 71 | ||
Set Membership: |
|