Search
Search

Slc25a54| Gene Symbol: Slc25a54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109640.1 | NP_001103110.1 | 1 - 2 | 257 | 63 | |
| XM_008761423.1 | XP_008759645.1 | 1 - 2 | 278 | 63 | ||
Set Membership: |
|