Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518638_m1
          See other FUBP1 GE Assays ›
          Gene Symbol:
          Fubp1
          Gene Name:
          far upstream element (FUSE) binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 257425676 - 257452745 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518638_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fubp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037653.1 NP_001032742.1 18 - 19 1824 59
          XM_006233507.2 XP_006233569.1 19 - 20 1935 59
          XM_006233511.2 XP_006233573.1 19 - 20 1935 59
          XM_006233512.2 XP_006233574.1 19 - 20 1926 59
          XM_006233513.2 XP_006233575.1 18 - 19 1862 59
          XM_008761553.1 XP_008759775.1 19 - 20 1949 59
          XM_008761554.1 XP_008759776.1 19 - 20 1926 59
          XM_008761555.1 XP_008759777.1 18 - 19 1871 59
          XM_008761556.1 XP_008759778.1 18 - 19 1868 59
          XM_008761557.1 XP_008759779.1 18 - 19 1859 59
          GenBank mRNA BC107942.1 - 18 - 19 1824 7389449 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002407.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fubp1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of gene expression
          positive regulation of transcription from RNA polymerase II promoter
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P694872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P694872 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P694872 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P694872 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1059463 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P694872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940673 - -
          Affymetrix Rat Gene 1.1 ST Array 10940673 - -
          Rat Genome 230 2.0 Array 1383287_at - -
          Rat Genome 230 2.0 Array 1383287_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200016899.rn.1 TC0200002407.rn.1 RTA TC
          Rat Expression Set 230 1383287_PM_at - -
          Rat Expression Set 230 1383287_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940673 - -
          Rat Genome U34 Set rc_AA997779_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361276 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline