Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518752_m1
          See other SSU72 GE Assays ›
          Gene Symbol:
          Ssu72
          Gene Name:
          SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae)
          Gene Aliases:
          RGD1307854
          Chromosome Location:
          Chr.5: 173152964 - 173182720 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518752_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ssu72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025657.1 NP_001020828.1 1 - 2 344 58
          GenBank mRNA BC099142.1 - 1 - 2 344 7300601 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002071.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ssu72

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA polyadenylation
          biological_process
          dephosphorylation of RNA polymerase II C-terminal domain
          molecular_function
          CTD phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144068 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039853 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P660334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61891 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874694 - -
          Affymetrix Rat Gene 1.0 ST Array 10874695 - -
          Affymetrix Rat Gene 1.0 ST Array 10874697 - -
          Affymetrix Rat Gene 1.0 ST Array 10874696 - -
          Affymetrix Rat Gene 1.0 ST Array 10874698 - -
          Affymetrix Rat Gene 1.1 ST Array 10874693 - -
          Rat Genome 230 2.0 Array 1371768_at - -
          Rat Genome 230 2.0 Array 1377297_PM_at - -
          Rat Genome 230 2.0 Array 1377297_at - -
          Rat Genome 230 2.0 Array 1371768_PM_at - -
          Affymetrix HT Rat Focus Array 1371768_at - -
          Affymetrix HT Rat Focus Array 1371768_PM_at - -
          Affymetrix HT Rat Focus Array 1377297_at - -
          Affymetrix HT Rat Focus Array 1377297_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500015752.rn.1 TC0500002071.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500015753.rn.1 TC0500002071.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500015755.rn.1 TC0500002071.rn.1 RTA TC
          Rat Expression Set 230 1377297_PM_at - -
          Rat Expression Set 230 1371768_PM_at - -
          Rat Expression Set 230 1371768_at - -
          Rat Expression Set 230 1377297_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874693 - -
          Rat Genome U34 Set rc_AI172260_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22204345 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline