Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518878_m1
          See other TMEM57 GE Assays ›
          Gene Symbol:
          Tmem57
          Gene Name:
          transmembrane protein 57
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 153122330 - 153184940 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518878_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025699.2 NP_001020870.2 9 - 10 1797 74
          XM_008764240.1 XP_008762462.1 8 - 9 1993 74
          XM_008764241.1 XP_008762463.1 9 - 10 1749 74
          GenBank mRNA AY845021.1 - 9 - 10 1797 74
          BC097999.1 - 9 - 10 1723 7453136 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003822.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem57

          Gene Ontology Categories:

          Function(s)

          brain development

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P999889 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P161692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P161692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P161692 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P161692 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P999889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P999889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161692 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P518016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83126 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880568 - -
          Affymetrix Rat Gene 1.0 ST Array 10880569 - -
          Affymetrix Rat Gene 1.0 ST Array 10880567 - -
          Affymetrix Rat Gene 1.0 ST Array 10880570 - -
          Affymetrix Rat Gene 1.0 ST Array 10880572 - -
          Affymetrix Rat Gene 1.0 ST Array 10880574 - -
          Affymetrix Rat Gene 1.0 ST Array 10880571 - -
          Affymetrix Rat Gene 1.0 ST Array 10880566 - -
          Affymetrix Rat Gene 1.0 ST Array 10880565 - -
          Affymetrix Rat Gene 1.0 ST Array 10880573 - -
          Affymetrix Rat Gene 1.1 ST Array 10880564 - -
          Rat Genome 230 2.0 Array 1393136_PM_at - -
          Rat Genome 230 2.0 Array 1393136_at - -
          Affymetrix HT Rat Focus Array 1393136_PM_at - -
          Affymetrix HT Rat Focus Array 1393136_at - -
          Affymetrix Rat Clariom S Assay PSR0500028719.rn.1 TC0500003822.rn.1 RTA TC
          Rat Expression Set 230 1393136_PM_at - -
          Rat Expression Set 230 1393136_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880564 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650041 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline