Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518925_m1
          See other ATAD3A GE Assays ›
          Gene Symbol:
          Atad3a
          Gene Name:
          ATPase family, AAA domain containing 3A
          Gene Aliases:
          Atad3
          Chromosome Location:
          Chr.5: 173189590 - 173209768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518925_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atad3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034922.1 NP_001030094.1 15 - 16 1658 67
          GenBank mRNA BC105762.1 - 15 - 16 1658 7929859 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004208.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atad3a

          Gene Ontology Categories:

          Function(s) Process(es)

          cell growth
          negative regulation of apoptotic process
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P525519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P525519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P525519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882205 - -
          Affymetrix Rat Gene 1.0 ST Array 10882220 - -
          Affymetrix Rat Gene 1.0 ST Array 10882214 - -
          Affymetrix Rat Gene 1.0 ST Array 10882215 - -
          Affymetrix Rat Gene 1.0 ST Array 10882219 - -
          Affymetrix Rat Gene 1.0 ST Array 10882209 - -
          Affymetrix Rat Gene 1.0 ST Array 10882206 - -
          Affymetrix Rat Gene 1.0 ST Array 10882207 - -
          Affymetrix Rat Gene 1.0 ST Array 10882216 - -
          Affymetrix Rat Gene 1.0 ST Array 10882211 - -
          Affymetrix Rat Gene 1.0 ST Array 10882203 - -
          Affymetrix Rat Gene 1.0 ST Array 10882210 - -
          Affymetrix Rat Gene 1.0 ST Array 10882202 - -
          Affymetrix Rat Gene 1.0 ST Array 10882213 - -
          Affymetrix Rat Gene 1.0 ST Array 10882212 - -
          Affymetrix Rat Gene 1.0 ST Array 10882208 - -
          Affymetrix Rat Gene 1.0 ST Array 10882218 - -
          Affymetrix Rat Gene 1.0 ST Array 10882204 - -
          Affymetrix Rat Gene 1.1 ST Array 10882201 - -
          Rat Genome 230 2.0 Array 1371980_at - -
          Rat Genome 230 2.0 Array 1371980_PM_at - -
          Affymetrix HT Rat Focus Array 1371980_PM_at - -
          Affymetrix HT Rat Focus Array 1371980_at - -
          Affymetrix Rat Clariom S Assay PSR0500032140.rn.1 TC0500004208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500032141.rn.1 TC0500004208.rn.1 RTA TC
          Rat Expression Set 230 1371980_at - -
          Rat Expression Set 230 1371980_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882201 - -
          Rat Genome U34 Set rc_AI103458_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21248459 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375075 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline