Search
Search

LOC500567| Gene Symbol: LOC500567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024348.1 | NP_001019519.1 | 1 - 2 | 268 | 113 | |
| GenBank mRNA | BC087049.1 | - | 1 - 2 | 268 | 7112813 | 113 |
Set Membership: |
|