Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518961_m1
          See other LDLRAP1 GE Assays ›
          Gene Symbol:
          Ldlrap1
          Gene Name:
          low density lipoprotein receptor adaptor protein 1
          Gene Aliases:
          RGD1563417
          Chromosome Location:
          Chr.5: 152964294 - 152987286 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518961_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ldlrap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109271.1 NP_001102741.1 7 - 8 817 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003820.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          biological_process
          positive regulation of signal transduction
          receptor internalization
          cholesterol homeostasis
          amyloid precursor protein metabolic process
          regulation of protein binding
          positive regulation of receptor-mediated endocytosis
          regulation of establishment of protein localization to plasma membrane
          receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport
          beta-amyloid binding
          phosphotyrosine binding
          molecular_function
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          receptor signaling complex scaffold activity
          clathrin binding
          signaling adaptor activity
          AP-2 adaptor complex binding
          clathrin adaptor activity
          low-density lipoprotein particle receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023088 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023088 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023088 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023083 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P274658 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880558 - -
          Affymetrix Rat Gene 1.0 ST Array 10880561 - -
          Affymetrix Rat Gene 1.0 ST Array 10880555 - -
          Affymetrix Rat Gene 1.0 ST Array 10880557 - -
          Affymetrix Rat Gene 1.0 ST Array 10880559 - -
          Affymetrix Rat Gene 1.0 ST Array 10880554 - -
          Affymetrix Rat Gene 1.0 ST Array 10880553 - -
          Affymetrix Rat Gene 1.0 ST Array 10880556 - -
          Affymetrix Rat Gene 1.0 ST Array 10880560 - -
          Affymetrix Rat Gene 1.1 ST Array 10880552 - -
          Rat Genome 230 2.0 Array 1395967_PM_at - -
          Rat Genome 230 2.0 Array 1395967_at - -
          Rat Genome 230 2.0 Array 1381672_at - -
          Rat Genome 230 2.0 Array 1381672_PM_at - -
          Affymetrix HT Rat Focus Array 1381672_PM_at - -
          Affymetrix HT Rat Focus Array 1381672_at - -
          Affymetrix Rat Clariom S Assay PSR0500028681.rn.1 TC0500003820.rn.1 RTA TC
          Rat Expression Set 230 1395967_PM_at - -
          Rat Expression Set 230 1381672_at - -
          Rat Expression Set 230 1395967_at - -
          Rat Expression Set 230 1381672_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880552 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21583964 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367181 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline