Search
Search

RGD1560436| Gene Symbol: RGD1560436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109264.1 | NP_001102734.1 | 3 - 4 | 801 | 65 | |
| XM_006238898.2 | XP_006238960.1 | 3 - 4 | 848 | 65 | ||
| XM_006238900.2 | XP_006238962.1 | 4 - 5 | 749 | 65 | ||
| XM_008764065.1 | XP_008762287.1 | 3 - 4 | 851 | 65 | ||
Set Membership: |
|