Search
Search

Tmem39b| Gene Symbol: Tmem39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014192.1 | NP_001014214.1 | 3 - 4 | 454 | 110 | |
| GenBank mRNA | BC082025.1 | - | 3 - 4 | 454 | 7129968 | 110 |
Set Membership: |
|