Search
Search
| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017209.1 | NP_058905.1 | 4 - 5 | 569 | 65 | |
| XM_006239009.2 | XP_006239071.1 | 5 - 6 | 688 | 65 | ||
| GenBank mRNA | AB015724.1 | - | 4 - 5 | 569 | 65 | |
| FQ210707.1 | - | 4 - 5 | 565 | 65 | ||
Set Membership: |
|