Search
Search

LOC682102| Gene Symbol: LOC682102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134732.1 | NP_001128204.1 | 3 - 4 | 1404 | 73 | |
| GenBank mRNA | BC168197.1 | - | 2 - 3 | 711 | 7112960 | 73 |
Set Membership: |
|