Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520391_m1
          See other TP73 GE Assays ›
          Gene Symbol:
          Tp73
          Gene Name:
          tumor protein p73
          Gene Aliases:
          P73, Trp73
          Chromosome Location:
          Chr.5: 171355876 - 171415354 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tp73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108696.1 NP_001102166.1 9 - 10 1213 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004172.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          kidney development
          release of cytochrome c from mitochondria
          transcription from RNA polymerase II promoter
          inflammatory response
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          cell cycle arrest
          regulation of mitotic cell cycle
          post-embryonic development
          response to X-ray
          response to organonitrogen compound
          response to gamma radiation
          regulation of gene expression
          hippocampus development
          forebrain development
          mitotic G1 DNA damage checkpoint
          cerebrospinal fluid secretion
          cellular response to UV
          response to drug
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of apoptotic process
          negative regulation of JUN kinase activity
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of cell size
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract morphogenesis
          neuron development
          positive regulation of oligodendrocyte differentiation
          protein tetramerization
          negative regulation of cardiac muscle cell proliferation
          positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor binding
          protein kinase binding
          identical protein binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079326 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079326 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P159382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91472 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72228 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36316 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882052 - -
          Affymetrix Rat Gene 1.0 ST Array 10882056 - -
          Affymetrix Rat Gene 1.0 ST Array 10882051 - -
          Affymetrix Rat Gene 1.0 ST Array 10882058 - -
          Affymetrix Rat Gene 1.0 ST Array 10882059 - -
          Affymetrix Rat Gene 1.0 ST Array 10882062 - -
          Affymetrix Rat Gene 1.0 ST Array 10882054 - -
          Affymetrix Rat Gene 1.0 ST Array 10882053 - -
          Affymetrix Rat Gene 1.0 ST Array 10882055 - -
          Affymetrix Rat Gene 1.0 ST Array 10882060 - -
          Affymetrix Rat Gene 1.0 ST Array 10882061 - -
          Affymetrix Rat Gene 1.0 ST Array 10882063 - -
          Affymetrix Rat Gene 1.0 ST Array 10882064 - -
          Affymetrix Rat Gene 1.0 ST Array 10882057 - -
          Affymetrix Rat Gene 1.1 ST Array 10882050 - -
          Affymetrix Rat Clariom S Assay PSR0500031826.rn.1 TC0500004172.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10882050 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22176515 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline