Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520757_m1
          See other SMPDL3B GE Assays ›
          Gene Symbol:
          Smpdl3b
          Gene Name:
          sphingomyelin phosphodiesterase, acid-like 3B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 150925150 - 150946994 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520757_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smpdl3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025737.1 NP_001020908.1 7 - 8 1115 85
          GenBank mRNA BC097983.1 - 7 - 8 1115 7443444 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003759.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smpdl3b

          Panther Classification:

          Molecular Function -

          hydrolase phosphodiesterase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingomyelin catabolic process
          sphingomyelin phosphodiesterase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30555 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25381 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872677 - -
          Affymetrix Rat Gene 1.0 ST Array 10872675 - -
          Affymetrix Rat Gene 1.0 ST Array 10872683 - -
          Affymetrix Rat Gene 1.0 ST Array 10872674 - -
          Affymetrix Rat Gene 1.0 ST Array 10872676 - -
          Affymetrix Rat Gene 1.0 ST Array 10872672 - -
          Affymetrix Rat Gene 1.0 ST Array 10872673 - -
          Affymetrix Rat Gene 1.0 ST Array 10880341 - -
          Affymetrix Rat Gene 1.0 ST Array 10880340 - -
          Affymetrix Rat Gene 1.0 ST Array 10872682 - -
          Affymetrix Rat Gene 1.0 ST Array 10872687 - -
          Affymetrix Rat Gene 1.0 ST Array 10872678 - -
          Affymetrix Rat Gene 1.1 ST Array 10872671 - -
          Affymetrix Rat Gene 1.1 ST Array 10872681 - -
          Affymetrix Rat Gene 1.1 ST Array 10880339 - -
          Affymetrix Rat Gene 1.1 ST Array 10872686 - -
          Rat Genome 230 2.0 Array 1376642_PM_at - -
          Rat Genome 230 2.0 Array 1373850_at - -
          Rat Genome 230 2.0 Array 1373850_PM_at - -
          Rat Genome 230 2.0 Array 1376642_at - -
          Affymetrix HT Rat Focus Array 1373850_at - -
          Affymetrix HT Rat Focus Array 1373850_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028072.rn.1 TC0500003759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028071.rn.1 TC0500003759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028073.rn.1 TC0500003759.rn.1 RTA TC
          Rat Expression Set 230 1373850_at - -
          Rat Expression Set 230 1376642_at - -
          Rat Expression Set 230 1373850_PM_at - -
          Rat Expression Set 230 1376642_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872686 - -
          Affymetrix Rat Gene 1.0 ST Array 10872671 - -
          Affymetrix Rat Gene 1.0 ST Array 10872681 - -
          Rat Genome U34 Set rc_AA997386_at - -
          Rat Genome U34 Set rc_AI014114_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline