Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520952_m1
          See other MEGF6 GE Assays ›
          Gene Symbol:
          Megf6
          Gene Name:
          multiple EGF-like-domains 6
          Gene Aliases:
          Egfl3
          Chromosome Location:
          Chr.5: 171588901 - 171620947 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520952_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Megf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022955.1 NP_075244.1 4894 63
          GenBank mRNA AB011532.1 - 4894 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002028.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Megf6

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Process(es)

          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P607202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874474 - -
          Affymetrix Rat Gene 1.0 ST Array 10874460 - -
          Affymetrix Rat Gene 1.0 ST Array 10874490 - -
          Affymetrix Rat Gene 1.0 ST Array 10874461 - -
          Affymetrix Rat Gene 1.0 ST Array 10874466 - -
          Affymetrix Rat Gene 1.0 ST Array 10874469 - -
          Affymetrix Rat Gene 1.0 ST Array 10874479 - -
          Affymetrix Rat Gene 1.0 ST Array 10874465 - -
          Affymetrix Rat Gene 1.0 ST Array 10874489 - -
          Affymetrix Rat Gene 1.0 ST Array 10874475 - -
          Affymetrix Rat Gene 1.0 ST Array 10874476 - -
          Affymetrix Rat Gene 1.0 ST Array 10874472 - -
          Affymetrix Rat Gene 1.0 ST Array 10874485 - -
          Affymetrix Rat Gene 1.0 ST Array 10874462 - -
          Affymetrix Rat Gene 1.0 ST Array 10874473 - -
          Affymetrix Rat Gene 1.0 ST Array 10874470 - -
          Affymetrix Rat Gene 1.0 ST Array 10874480 - -
          Affymetrix Rat Gene 1.0 ST Array 10874478 - -
          Affymetrix Rat Gene 1.0 ST Array 10874487 - -
          Affymetrix Rat Gene 1.0 ST Array 10874468 - -
          Affymetrix Rat Gene 1.0 ST Array 10874464 - -
          Affymetrix Rat Gene 1.0 ST Array 10874463 - -
          Affymetrix Rat Gene 1.0 ST Array 10874467 - -
          Affymetrix Rat Gene 1.0 ST Array 10874486 - -
          Affymetrix Rat Gene 1.0 ST Array 10874488 - -
          Affymetrix Rat Gene 1.0 ST Array 10874477 - -
          Affymetrix Rat Gene 1.0 ST Array 10874471 - -
          Affymetrix Rat Gene 1.0 ST Array 10874482 - -
          Affymetrix Rat Gene 1.0 ST Array 10874483 - -
          Affymetrix Rat Gene 1.0 ST Array 10874481 - -
          Affymetrix Rat Gene 1.0 ST Array 10874484 - -
          Affymetrix Rat Gene 1.1 ST Array 10874458 - -
          Rat Genome 230 2.0 Array 1387439_at - -
          Rat Genome 230 2.0 Array 1387439_PM_at - -
          Affymetrix HT Rat Focus Array 1387439_at - -
          Affymetrix HT Rat Focus Array 1387439_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500015368.rn.1 TC0500002028.rn.1 RTA TC
          Rat Expression Set 230 1387439_at - -
          Rat Expression Set 230 1387439_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874458 - -
          Rat Genome U34 Set AB011532_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21712253 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline