Search
Search
| Gene Symbol: Cetn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229519.1 | XP_006229581.1 | 2 - 3 | 301 | 146 | |
| XM_215222.7 | XP_215222.5 | 2 - 3 | 437 | 146 | ||
| GenBank mRNA | FQ219097.1 | - | 2 - 3 | 195 | 146 | |
Set Membership: |
|