Search
Search
| Gene Symbol: Dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013076.1 | NP_001013094.1 | 5 - 6 | 1178 | 148 | |
| XM_006233488.1 | XP_006233550.1 | 4 - 5 | 1160 | 148 | ||
| XM_008761550.1 | XP_008759772.1 | 2 - 3 | 1545 | 148 | ||
| GenBank mRNA | BC083638.1 | - | 5 - 6 | 1178 | 7134832 | 148 |
Set Membership: |
|