Search
Search
| Gene Symbol: Rnpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100810.1 | NP_001094280.1 | 1 - 2 | 233 | 98 | |
| XM_006233211.2 | XP_006233273.1 | 1 - 2 | 611 | 98 | ||
| XM_008761424.1 | XP_008759646.1 | 1 - 2 | 611 | 98 | ||
| XM_008761425.1 | XP_008759647.1 | 1 - 2 | 612 | 98 | ||
| GenBank mRNA | BC166571.1 | - | 1 - 2 | 233 | 7320328 | 98 |
Set Membership: |
|