Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01521539_m1
          See other SPATA1 GE Assays ›
          Gene Symbol:
          Spata1
          Gene Name:
          spermatogenesis associated 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 252313293 - 252359798 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01521539_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014177.1 NP_001014199.1 12 - 13 1426 150
          XM_008761517.1 XP_008759739.1 12 - 13 1715 150
          GenBank mRNA BC079224.1 - 12 - 13 1426 7132232 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004870.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata1

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109784 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109784 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991456 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827269 - -
          Affymetrix Rat Gene 1.0 ST Array 10827272 - -
          Affymetrix Rat Gene 1.0 ST Array 10827275 - -
          Affymetrix Rat Gene 1.0 ST Array 10827266 - -
          Affymetrix Rat Gene 1.0 ST Array 10819731 - -
          Affymetrix Rat Gene 1.0 ST Array 10827274 - -
          Affymetrix Rat Gene 1.0 ST Array 10827278 - -
          Affymetrix Rat Gene 1.0 ST Array 10827271 - -
          Affymetrix Rat Gene 1.0 ST Array 10827277 - -
          Affymetrix Rat Gene 1.0 ST Array 10827273 - -
          Affymetrix Rat Gene 1.0 ST Array 10827265 - -
          Affymetrix Rat Gene 1.0 ST Array 10827276 - -
          Affymetrix Rat Gene 1.0 ST Array 10827267 - -
          Affymetrix Rat Gene 1.0 ST Array 10827268 - -
          Affymetrix Rat Gene 1.0 ST Array 10827270 - -
          Affymetrix Rat Gene 1.1 ST Array 10819726 - -
          Affymetrix Rat Gene 1.1 ST Array 10827264 - -
          Rat Genome 230 2.0 Array 1393724_at - -
          Rat Genome 230 2.0 Array 1393724_PM_at - -
          Affymetrix HT Rat Focus Array 1393724_at - -
          Affymetrix HT Rat Focus Array 1393724_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200033679.rn.1 TC0200004870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033677.rn.1 TC0200004870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033676.rn.1 TC0200004870.rn.1 RTA TC
          Rat Expression Set 230 1393724_at - -
          Rat Expression Set 230 1393724_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10827264 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22377285 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline