Search
Search
| Gene Symbol: Dph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017449.1 | NP_001017449.1 | 4 - 5 | 693 | 95 | |
| XM_006233214.1 | XP_006233276.1 | 4 - 5 | 506 | 95 | ||
| XM_008761454.1 | XP_008759676.1 | 3 - 4 | 554 | 95 | ||
| XM_008761455.1 | XP_008759677.1 | 3 - 4 | 555 | 95 | ||
| GenBank mRNA | BC092598.1 | - | 4 - 5 | 693 | 7321341 | 95 |
Set Membership: |
|