Search
Search
| Gene Symbol: Psrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044302.1 | NP_001037767.1 | 6 - 7 | 950 | 82 | |
| XM_006233176.2 | XP_006233238.1 | 6 - 7 | 1160 | 82 | ||
| XM_006233177.2 | XP_006233239.1 | 6 - 7 | 991 | 82 | ||
| XM_006233178.2 | XP_006233240.1 | 6 - 7 | 1160 | 82 | ||
| XM_006233179.2 | XP_006233241.1 | 6 - 7 | 991 | 82 | ||
| GenBank mRNA | BC105874.1 | - | 6 - 7 | 950 | 7131142 | 82 |
Set Membership: |
|