Search
Search

RGD1309110| Gene Symbol: RGD1309110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134512.1 | NP_001127984.1 | 3 - 4 | 302 | 67 | |
| XM_006233113.2 | XP_006233175.1 | 4 - 5 | 453 | 67 | ||
Set Membership: |
|