Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522048_m1
          See other CRYZ GE Assays ›
          Gene Symbol:
          Cryz
          Gene Name:
          crystallin, zeta (quinone reductase)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 260884318 - 260911827 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522048_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cryz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012183.2 NP_001012183.1 6 - 7 703 88
          XM_006233520.2 XP_006233582.1 7 - 8 939 88
          GenBank mRNA BC078927.1 - 6 - 7 705 7106704 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002442.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cryz

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          xenobiotic catabolic process
          protein homotetramerization
          oxidation-reduction process
          mRNA 3'-UTR binding
          NADPH:quinone reductase activity
          zinc ion binding
          NADPH binding
          NADH binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109207 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23049 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23049 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23049 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56111 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819836 - -
          Affymetrix Rat Gene 1.0 ST Array 10819829 - -
          Affymetrix Rat Gene 1.0 ST Array 10819837 - -
          Affymetrix Rat Gene 1.0 ST Array 10819832 - -
          Affymetrix Rat Gene 1.0 ST Array 10819834 - -
          Affymetrix Rat Gene 1.0 ST Array 10819835 - -
          Affymetrix Rat Gene 1.0 ST Array 10819830 - -
          Affymetrix Rat Gene 1.0 ST Array 10819831 - -
          Affymetrix Rat Gene 1.0 ST Array 10819833 - -
          Affymetrix Rat Gene 1.1 ST Array 10819828 - -
          Rat Genome 230 2.0 Array 1374628_at - -
          Rat Genome 230 2.0 Array 1380393_PM_at - -
          Rat Genome 230 2.0 Array 1380393_at - -
          Rat Genome 230 2.0 Array 1374628_PM_at - -
          Affymetrix HT Rat Focus Array 1380393_at - -
          Affymetrix HT Rat Focus Array 1374628_PM_at - -
          Affymetrix HT Rat Focus Array 1374628_at - -
          Affymetrix HT Rat Focus Array 1380393_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200017107.rn.1 TC0200002442.rn.1 RTA TC
          Rat Expression Set 230 1374628_PM_at - -
          Rat Expression Set 230 1374628_at - -
          Rat Expression Set 230 1380393_PM_at - -
          Rat Expression Set 230 1380393_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819828 - -
          Rat Genome U34 Set rc_AI059338_at - -
          Rat Genome U34 Set rc_AI232098_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21424827 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline