Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522160_m1
          See other AMY1A GE Assays ›
          Gene Symbol:
          Amy1a
          Gene Name:
          amylase, alpha 1A (salivary)
          Gene Aliases:
          Amy1
          Chromosome Location:
          Chr.2: 216428709 - 216443518 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522160_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amy1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010970.1 NP_001010970.1 4 - 5 585 63
          GenBank mRNA AB057450.1 - 4 - 5 585 63
          BC088228.1 - 3 - 4 528 7323054 63
          FQ219213.1 - 4 - 5 914 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004513.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amy1a

          Panther Classification:

          Molecular Function -

          amylase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          alpha-amylase activity
          amylase activity
          cation binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P435713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P435713 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P435713 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P435713 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P435713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826215 - -
          Affymetrix Rat Gene 1.0 ST Array 10826176 - -
          Affymetrix Rat Gene 1.0 ST Array 10826214 - -
          Affymetrix Rat Gene 1.0 ST Array 10826185 - -
          Affymetrix Rat Gene 1.0 ST Array 10826213 - -
          Affymetrix Rat Gene 1.0 ST Array 10826188 - -
          Affymetrix Rat Gene 1.0 ST Array 10826207 - -
          Affymetrix Rat Gene 1.0 ST Array 10826186 - -
          Affymetrix Rat Gene 1.0 ST Array 10826210 - -
          Affymetrix Rat Gene 1.0 ST Array 10826191 - -
          Affymetrix Rat Gene 1.0 ST Array 10826212 - -
          Affymetrix Rat Gene 1.0 ST Array 10826177 - -
          Affymetrix Rat Gene 1.0 ST Array 10826208 - -
          Affymetrix Rat Gene 1.0 ST Array 10826180 - -
          Affymetrix Rat Gene 1.0 ST Array 10826209 - -
          Affymetrix Rat Gene 1.0 ST Array 10826211 - -
          Affymetrix Rat Gene 1.1 ST Array 10826184 - -
          Affymetrix Rat Gene 1.1 ST Array 10826206 - -
          Affymetrix Rat Gene 1.1 ST Array 10826175 - -
          Rat Genome 230 2.0 Array 1370359_PM_at - -
          Rat Genome 230 2.0 Array 1369502_a_at - -
          Rat Genome 230 2.0 Array 1370359_at - -
          Rat Genome 230 2.0 Array 1369502_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370359_PM_at - -
          Affymetrix HT Rat Focus Array 1370359_at - -
          Affymetrix Rat Clariom S Assay PSR0200031410.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031412.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031408.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031409.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031411.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031407.rn.1 TC0200004513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200031413.rn.1 TC0200004513.rn.1 RTA TC
          Rat Expression Set 230 1370359_at - -
          Rat Expression Set 230 1369502_a_at - -
          Rat Expression Set 230 1370359_PM_at - -
          Rat Expression Set 230 1369502_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826206 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22350146 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline