Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522267_m1
          See other LPPR4 GE Assays ›
          Gene Symbol:
          Lppr4
          Gene Name:
          lipid phosphate phosphatase-related protein type 4
          Gene Aliases:
          Prg-1, Prg1
          Chromosome Location:
          Chr.2: 220298239 - 220341863 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522267_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lppr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001508.2 NP_001001508.2 6 - 7 1559 95
          GenBank mRNA AF541280.1 - 6 - 7 1560 95
          AY266268.1 - 6 - 7 1560 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002057.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lppr4

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          axonogenesis
          phospholipid dephosphorylation
          inner ear development
          phosphatidate phosphatase activity
          lipid phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P196296 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140813 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196296 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196296 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P196296 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P196296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41111 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98432 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826396 - -
          Affymetrix Rat Gene 1.0 ST Array 10826402 - -
          Affymetrix Rat Gene 1.0 ST Array 10826401 - -
          Affymetrix Rat Gene 1.0 ST Array 10826397 - -
          Affymetrix Rat Gene 1.0 ST Array 10826400 - -
          Affymetrix Rat Gene 1.0 ST Array 10826394 - -
          Affymetrix Rat Gene 1.0 ST Array 10826395 - -
          Affymetrix Rat Gene 1.0 ST Array 10826399 - -
          Affymetrix Rat Gene 1.0 ST Array 10826393 - -
          Affymetrix Rat Gene 1.1 ST Array 10826392 - -
          Rat Genome 230 2.0 Array 1379374_at - -
          Rat Genome 230 2.0 Array 1385981_PM_at - -
          Rat Genome 230 2.0 Array 1385981_at - -
          Rat Genome 230 2.0 Array 1379374_PM_at - -
          Affymetrix HT Rat Focus Array 1379374_at - -
          Affymetrix HT Rat Focus Array 1385981_at - -
          Affymetrix HT Rat Focus Array 1385981_PM_at - -
          Affymetrix HT Rat Focus Array 1379374_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200014238.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014241.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014243.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014239.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014240.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014242.rn.1 TC0200002057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014244.rn.1 TC0200002057.rn.1 RTA TC
          Rat Expression Set 230 1379374_at - -
          Rat Expression Set 230 1379374_PM_at - -
          Rat Expression Set 230 1385981_at - -
          Rat Expression Set 230 1385981_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826392 - -
          Rat Genome U34 Set rc_AI228462_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20985086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline