Search
Search
| Gene Symbol: Csf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023981.4 | NP_076471.3 | 3 - 4 | 387 | 115 | |
| XM_006233187.1 | XP_006233249.1 | 3 - 4 | 258 | 115 | ||
| XM_008761426.1 | XP_008759648.1 | 3 - 4 | 258 | 115 | ||
| XM_008761427.1 | XP_008759649.1 | 3 - 4 | 258 | 115 | ||
| XM_008761428.1 | XP_008759650.1 | 3 - 4 | 258 | 115 | ||
| XM_008761429.1 | XP_008759651.1 | 3 - 4 | 258 | 115 | ||
| GenBank mRNA | AF514355.1 | - | 3 - 4 | 227 | 115 | |
| AF514356.1 | - | 3 - 4 | 227 | 115 | ||
| AF514357.1 | - | 3 - 4 | 237 | 115 | ||
| AF515736.1 | - | 3 - 4 | 325 | 115 | ||
| BC074007.1 | - | 3 - 4 | 387 | 7099454 | 115 | |
Set Membership: |
|