Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022294.1 | NP_071630.1 | 12 - 13 | 2056 | 87 | |
GenBank mRNA | AF192401.1 | - | 12 - 13 | 2056 | 87 | |
AF192402.1 | - | 12 - 13 | 2056 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |