Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522901_g1
          See other TMEM167B GE Assays ›
          Gene Symbol:
          Tmem167b
          Gene Name:
          transmembrane protein 167B
          Gene Aliases:
          RGD1564454
          Chromosome Location:
          Chr.2: 211348762 - 211352540 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522901_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem167b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135260.1 NP_001128732.1 2 - 3 335 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099269 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099264 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832916 - -
          Affymetrix Rat Gene 1.0 ST Array 10826080 - -
          Affymetrix Rat Gene 1.0 ST Array 10826081 - -
          Affymetrix Rat Gene 1.1 ST Array 10826079 - -
          Affymetrix Rat Gene 1.1 ST Array 10832915 - -
          Rat Genome 230 2.0 Array 1377964_at - -
          Rat Genome 230 2.0 Array 1377964_PM_at - -
          Rat Genome 230 2.0 Array 1393186_PM_at - -
          Rat Genome 230 2.0 Array 1393186_at - -
          Affymetrix Rat Clariom S Assay PSR0200031154.rn.1 TC0200004476.rn.1 RTA TC
          Rat Expression Set 230 1393186_at - -
          Rat Expression Set 230 1393186_PM_at - -
          Rat Expression Set 230 1377964_at - -
          Rat Expression Set 230 1377964_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826079 - -
          Rat Genome U34 Set rc_AI013181_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22371768 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353110 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline