Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522910_g1
          See other RPL34 GE Assays ›
          Gene Symbol:
          Rpl34
          Gene Name:
          ribosomal protein L34
          Gene Aliases:
          Rpl34l2
          Chromosome Location:
          Chr.2: 235850281 - 235867129 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522910_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108567.2 NP_001102037.1 2 - 3 97 103
          XM_006233314.2 XP_006233376.1 2 - 3 262 103
          XM_006233315.1 XP_006233377.1 1 - 2 85 103
          GenBank mRNA BC167113.1 - 2 - 3 97 6889959 103
          FQ211292.1 - 2 - 3 100 103
          FQ211369.1 - 2 - 3 99 103
          FQ221629.1 - 2 - 3 99 103
          FQ221933.1 - 2 - 3 99 103
          FQ223193.1 - 2 - 3 99 103
          FQ224761.1 - 2 - 3 105 103
          FQ229743.1 - 2 - 3 99 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873865 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047893 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1031524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826820 - -
          Affymetrix Rat Gene 1.0 ST Array 10826821 - -
          Affymetrix Rat Gene 1.0 ST Array 10826819 - -
          Affymetrix Rat Gene 1.0 ST Array 10826822 - -
          Affymetrix Rat Gene 1.0 ST Array 10796417 - -
          Affymetrix Rat Gene 1.1 ST Array 10826818 - -
          Affymetrix Rat Gene 1.1 ST Array 10796416 - -
          Rat Genome 230 2.0 Array 1371761_at - -
          Rat Genome 230 2.0 Array 1371761_PM_at - -
          Affymetrix HT Rat Focus Array 1371761_at - -
          Affymetrix HT Rat Focus Array 1371761_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200032664.rn.1 TC0200004691.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032669.rn.1 TC0200004691.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032665.rn.1 TC0200004691.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032667.rn.1 TC0200004691.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032668.rn.1 TC0200004691.rn.1 RTA TC
          Rat Expression Set 230 1371761_PM_at - -
          Rat Expression Set 230 1371761_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796416 - -
          Affymetrix Rat Gene 1.0 ST Array 10826818 - -
          Rat Genome U34 Set rc_AI013910_at - -
          Rat Genome U34 Set rc_AI230691_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650183 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline