Search
Search
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191850.1 | NP_001178779.1 | 9 - 10 | 633 | 71 | |
| XM_006233291.2 | XP_006233353.2 | 9 - 10 | 966 | 71 | ||
| XM_006233292.2 | XP_006233354.2 | 8 - 9 | 867 | 71 | ||
| GenBank mRNA | BC091336.1 | - | 2 - 3 | 181 | 71 | |
Set Membership: |
|