Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523645_m1
          See other DENND2D GE Assays ›
          Gene Symbol:
          Dennd2d
          Gene Name:
          DENN/MADD domain containing 2D
          Gene Aliases:
          RGD1305120
          Chromosome Location:
          Chr.2: 209097894 - 209116971 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523645_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dennd2d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107714.1 NP_001101184.1 5 - 6 578 77
          XM_006233114.2 XP_006233176.1 5 - 6 527 77
          XM_008761396.1 XP_008759618.1 4 - 5 974 77
          XM_008761397.1 XP_008759619.1 4 - 5 974 77
          XM_008761398.1 XP_008759620.1 5 - 6 527 77
          XM_008761399.1 XP_008759621.1 5 - 6 611 77
          GenBank mRNA BC158880.1 - 5 - 6 514 7936253 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001945.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          Rab guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P465144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P465144 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P465144 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P465144 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P465144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818207 - -
          Affymetrix Rat Gene 1.0 ST Array 10818216 - -
          Affymetrix Rat Gene 1.0 ST Array 10818213 - -
          Affymetrix Rat Gene 1.0 ST Array 10818208 - -
          Affymetrix Rat Gene 1.0 ST Array 10818212 - -
          Affymetrix Rat Gene 1.0 ST Array 10818215 - -
          Affymetrix Rat Gene 1.0 ST Array 10818217 - -
          Affymetrix Rat Gene 1.0 ST Array 10818210 - -
          Affymetrix Rat Gene 1.0 ST Array 10818205 - -
          Affymetrix Rat Gene 1.0 ST Array 10818209 - -
          Affymetrix Rat Gene 1.0 ST Array 10818211 - -
          Affymetrix Rat Gene 1.0 ST Array 10818214 - -
          Affymetrix Rat Gene 1.1 ST Array 10818204 - -
          Rat Genome 230 2.0 Array 1380021_at - -
          Rat Genome 230 2.0 Array 1380021_PM_at - -
          Affymetrix HT Rat Focus Array 1380021_at - -
          Affymetrix HT Rat Focus Array 1380021_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200013389.rn.1 TC0200001945.rn.1 RTA TC
          Rat Expression Set 230 1380021_PM_at - -
          Rat Expression Set 230 1380021_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818204 - -
          Rat Genome U34 Set rc_AI059486_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21933611 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline